Assay Details
| Gene Symbol: abt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381367.1 | NP_001368466.1 | 27 - 28 | 4882 | 87 | |
| NM_058548.7 | NP_490949.3 | 27 - 28 | 4993 | 87 | ||
More Information
Set Membership: |
|

abt-2
Genomic Map