Assay Details
| Gene Symbol: abt-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381367.1 | NP_001368466.1 | 28 - 29 | 5009 | 105 | |
| NM_058548.7 | NP_490949.3 | 28 - 29 | 5120 | 105 | ||
More Information
Set Membership: |
|

abt-2
Genomic Map