Assay Details
| Gene Symbol: aat-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058601.6 | NP_491002.2 | 8 - 9 | 1366 | 81 | |
| NR_131358.1 | 8 - 9 | 1352 | 81 | |||
| NR_131359.1 | 7 - 8 | 1409 | 81 | |||
| NR_131780.1 | 8 - 9 | 1344 | 81 | |||
More Information
Set Membership: |
|

aat-5
Genomic Map