Assay Details
| Gene Symbol: rpl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381533.1 | NP_001368007.1 | 1 - 2 | 291 | 76 | |
| NM_058660.6 | NP_491061.1 | 2 - 3 | 480 | 76 | ||
| NR_002353.1 | 1 - 2 | 324 | 76 | |||
| NR_002354.1 | 2 - 3 | 336 | 76 | |||
More Information
Set Membership: |
|

rpl-1
Genomic Map