Assay Details
| Gene Symbol: arx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058665.7 | NP_491066.1 | 3 - 4 | 364 | 61 | |
| Gene Symbol: dnc-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381537.1 | NP_001368008.1 | 3 - 4 | 364 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381537.1 transcripts.
Set Membership: |
|

arx-1
Genomic Map