Assay Details
Gene Symbol: nxt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129079.3 | NP_001122551.1 | 3 - 4 | 421 | 85 | |
NM_058676.5 | NP_491077.1 | 3 - 4 | 480 | 85 | ||
GenBank mRNA | AF156960.1 | - | 3 - 4 | 376 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
