Assay Details
Gene Symbol: pign-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001262367.2 | NP_001249296.1 | 12 - 13 | 2601 | 123 | |
NM_001262368.4 | NP_001249297.1 | 12 - 13 | 4116 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
