Assay Details
Gene Symbol: pign-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001262367.2 | NP_001249296.1 | 5 - 6 | 1116 | 158 | |
NM_001262368.4 | NP_001249297.1 | 5 - 6 | 2649 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
