Assay Details
Gene Symbol: W05F2.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058746.6 | NP_491147.4 | 4 - 5 | 504 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: W05F2.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058746.6 | NP_491147.4 | 4 - 5 | 504 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() |