Assay Details
Gene Symbol: cdk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026366.3 | NP_001021537.2 | 5 - 6 | 732 | 107 | |
NM_001350362.3 | NP_001337286.1 | 5 - 6 | 749 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
