Assay Details
| Gene Symbol: crn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058767.8 | NP_491168.1 | 2 - 3 | 838 | 100 | |
| GenBank mRNA | AY303575.1 | - | 2 - 3 | 805 | 100 | |
More Information
Set Membership: |
|

crn-1
Genomic Map