Assay Details
| Gene Symbol: pcaf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058772.8 | NP_491173.1 | 10 - 11 | 2219 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pcaf-1| Gene Symbol: pcaf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058772.8 | NP_491173.1 | 10 - 11 | 2219 | 138 | |
Set Membership: |
|