Assay Details
| Gene Symbol: pcaf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058772.8 | NP_491173.1 | 3 - 4 | 468 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pcaf-1| Gene Symbol: pcaf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058772.8 | NP_491173.1 | 3 - 4 | 468 | 96 | |
Set Membership: |
|