Assay Details
Gene Symbol: abf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058852.7 | NP_491253.1 | 1 - 2 | 152 | 80 | |
GenBank mRNA | AB029809.1 | - | 1 - 2 | 328 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
