Assay Details
| Gene Symbol: unc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026325.2 | NP_001021496.1 | 24 - 25 | 6527 | 66 | |
| NM_001026327.4 | NP_001021498.1 | 8 - 9 | 2061 | 66 | ||
| NM_001026328.3 | NP_001021499.1 | 5 - 6 | 1624 | 66 | ||
| NM_001026329.6 | NP_001021500.1 | 6 - 7 | 1727 | 66 | ||
| NM_001262583.3 | NP_001249512.1 | 6 - 7 | 1720 | 66 | ||
| NM_001262584.2 | NP_001249513.1 | 8 - 9 | 2108 | 66 | ||
| NM_001375256.3 | NP_001361940.1 | 8 - 9 | 1855 | 66 | ||
| GenBank mRNA | AF048834.1 | - | 24 - 25 | 6527 | 66 | |
More Information
Set Membership: |
|

unc-73
Genomic Map