Assay Details
| Gene Symbol: nab-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379751.2 | NP_001367535.1 | 4 - 5 | 397 | 71 | |
| NM_001382891.1 | 2 - 3 | 588 | 71 | |||
| NM_001382892.1 | 4 - 5 | 394 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001382892.1 transcripts.
Set Membership: |
|

nab-1
Genomic Map