Assay Details
| Gene Symbol: xnp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025787.4 | NP_001020958.1 | 5 - 6 | 2050 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

xnp-1| Gene Symbol: xnp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025787.4 | NP_001020958.1 | 5 - 6 | 2050 | 137 | |
Set Membership: |
|