Assay Details
| Gene Symbol: paf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383031.1 | 10 - 11 | 2426 | 115 | ||
| GenBank mRNA | AF386744.1 | - | 5 - 6 | 580 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

paf-1| Gene Symbol: paf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383031.1 | 10 - 11 | 2426 | 115 | ||
| GenBank mRNA | AF386744.1 | - | 5 - 6 | 580 | 115 | |
Set Membership: |
|