Assay Details
Gene Symbol: chp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059118.6 | NP_491519.1 | 3 - 4 | 1445 | 75 | |
GenBank mRNA | AF192264.1 | - | 2 - 3 | 354 | 75 |
Gene Symbol: spe-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059117.3 | NP_491518.1 | 2 - 3 | 355 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_059117.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
