Assay Details
| Gene Symbol: pde-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059143.4 | NP_491544.3 | 11 - 12 | 1483 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pde-5| Gene Symbol: pde-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059143.4 | NP_491544.3 | 11 - 12 | 1483 | 104 | |
Set Membership: |
|