Assay Details
| Gene Symbol: mrpl-47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372756.1 | NP_001359789.1 | 3 - 4 | 473 | 73 | |
| NM_001380146.1 | NP_001367333.1 | 3 - 4 | 309 | 73 | ||
More Information
Set Membership: |
|

mrpl-47
Genomic Map