Assay Details
| Gene Symbol: let-363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059148.5 | NP_491549.2 | 30 - 31 | 7721 | 86 | |
| NM_059151.7 | NP_491552.2 | 30 - 31 | 7674 | 86 | ||
More Information
Set Membership: |
|

let-363
Genomic Map