Assay Details
| Gene Symbol: let-363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059148.5 | NP_491549.2 | 9 - 10 | 1743 | 62 | |
| NM_059151.7 | NP_491552.2 | 9 - 10 | 1705 | 62 | ||
More Information
Set Membership: |
|

let-363
Genomic Map