Assay Details
Gene Symbol: atp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026250.7 | NP_001021421.1 | 2 - 3 | 382 | 90 | |
NM_001383096.1 | 2 - 3 | 368 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
