Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 19 - 20 | 3284 | 127 | |
| NM_001318273.3 | NP_001305202.1 | 7 - 8 | 1043 | 127 | ||
| NM_001318274.3 | NP_001305203.1 | 3 - 4 | 284 | 127 | ||
| NM_001318275.3 | NP_001305204.1 | 1 - 2 | 110 | 127 | ||
| NM_001383115.1 | 20 - 21 | 3368 | 127 | |||
| NM_059236.5 | NP_491637.2 | 21 - 22 | 3407 | 127 | ||
| NM_059237.7 | NP_491638.1 | 3 - 4 | 873 | 127 | ||
More Information
Set Membership: |
|

tns-1
Genomic Map