Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 22 - 23 | 3643 | 94 | |
| NM_001318273.3 | NP_001305202.1 | 10 - 11 | 1402 | 94 | ||
| NM_001318274.3 | NP_001305203.1 | 6 - 7 | 643 | 94 | ||
| NM_001318275.3 | NP_001305204.1 | 4 - 5 | 469 | 94 | ||
| NM_001383115.1 | 23 - 24 | 3727 | 94 | |||
| NM_059236.5 | NP_491637.2 | 24 - 25 | 3766 | 94 | ||
| NM_059237.7 | NP_491638.1 | 6 - 7 | 1232 | 94 | ||
More Information
Set Membership: |
|

tns-1
Genomic Map