Assay Details
| Gene Symbol: cir-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059253.5 | NP_491654.1 | 2 - 3 | 496 | 93 | |
| GenBank mRNA | DQ645890.1 | - | 2 - 3 | 518 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cir-1| Gene Symbol: cir-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059253.5 | NP_491654.1 | 2 - 3 | 496 | 93 | |
| GenBank mRNA | DQ645890.1 | - | 2 - 3 | 518 | 93 | |
Set Membership: |
|