Assay Details
| Gene Symbol: hil-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059277.6 | NP_491678.1 | 1 - 2 | 273 | 107 | |
| GenBank mRNA | AF005372.1 | - | 1 - 2 | 245 | 107 | |
More Information
Set Membership: |
|

hil-5
Genomic Map