Assay Details
Gene Symbol: cpb-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059279.7 | NP_491680.2 | 4 - 5 | 1843 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
