Assay Details
| Gene Symbol: hmg-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059287.4 | NP_491688.1 | 3 - 4 | 1823 | 83 | |
| Gene Symbol: nhr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059288.8 | NP_491689.4 | 7 - 8 | 3998 | 83 | |
More Information
Set Membership: |
|

hmg-3
Genomic Map