Assay Details
Gene Symbol: ntl-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307221.3 | NP_001294150.1 | 10 - 11 | 1614 | 77 | |
NM_171388.6 | NP_741453.1 | 10 - 11 | 1613 | 77 | ||
NR_131578.1 | 9 - 10 | 1670 | 77 |
Gene Symbol: C49H3.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383131.1 | 10 - 11 | 1609 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383131.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
