Assay Details
| Gene Symbol: ntl-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307221.3 | NP_001294150.1 | 3 - 4 | 255 | 80 | |
| NM_001307222.3 | NP_001294151.1 | 3 - 4 | 273 | 80 | ||
| NM_001307223.3 | NP_001294152.1 | 3 - 4 | 254 | 80 | ||
| NM_171388.6 | NP_741453.1 | 3 - 4 | 248 | 80 | ||
| NR_131578.1 | 3 - 4 | 202 | 80 | |||
| Gene Symbol: C49H3.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383131.1 | 3 - 4 | 244 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383131.1 transcripts.
Set Membership: |
|

ntl-4
Genomic Map