Assay Details
| Gene Symbol: ttll-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380378.1 | NP_001367737.1 | 10 - 11 | 1409 | 147 | |
| NM_171405.7 | NP_741471.2 | 12 - 13 | 1706 | 147 | ||
More Information
Set Membership: |
|

ttll-11
Genomic Map