Assay Details
| Gene Symbol: ttll-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380378.1 | NP_001367737.1 | 13 - 14 | 1757 | 73 | |
| NM_171405.7 | NP_741471.2 | 15 - 16 | 2054 | 73 | ||
More Information
Set Membership: |
|

ttll-11
Genomic Map