Assay Details
| Gene Symbol: ttll-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380378.1 | NP_001367737.1 | 2 - 3 | 264 | 147 | |
| NM_171405.7 | NP_741471.2 | 4 - 5 | 561 | 147 | ||
More Information
Set Membership: |
|

ttll-11
Genomic Map