Assay Details
Gene Symbol: set-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060371.4 | NP_492772.1 | 5 - 6 | 1159 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
