Assay Details
| Gene Symbol: mrps-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060382.7 | NP_492783.1 | 4 - 5 | 1379 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrps-30| Gene Symbol: mrps-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060382.7 | NP_492783.1 | 4 - 5 | 1379 | 94 | |
Set Membership: |
|