Assay Details
| Gene Symbol: mec-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028946.6 | NP_001024117.2 | 22 - 23 | 4381 | 84 | |
| NM_001028947.3 | NP_001024118.1 | 19 - 20 | 4205 | 84 | ||
| NM_001028949.3 | NP_001024120.1 | 13 - 14 | 3155 | 84 | ||
| NM_001373047.2 | NP_001359916.1 | 20 - 21 | 4321 | 84 | ||
| NM_001380680.1 | NP_001367617.1 | 21 - 22 | 4361 | 84 | ||
| NM_001380681.1 | NP_001367615.1 | 21 - 22 | 4352 | 84 | ||
| NM_171477.7 | NP_741559.3 | 22 - 23 | 4417 | 84 | ||
More Information
Set Membership: |
|

mec-1
Genomic Map