Assay Details
| Gene Symbol: mec-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028946.6 | NP_001024117.2 | 3 - 4 | 287 | 72 | |
| NM_001373047.2 | NP_001359916.1 | 2 - 3 | 136 | 72 | ||
| NM_001380680.1 | NP_001367617.1 | 2 - 3 | 234 | 72 | ||
| NM_001380681.1 | NP_001367615.1 | 2 - 3 | 234 | 72 | ||
| NM_001380682.1 | NP_001367616.1 | 2 - 3 | 234 | 72 | ||
| NM_001380683.1 | NP_001367618.1 | 2 - 3 | 234 | 72 | ||
| NM_001380684.1 | NP_001367619.1 | 2 - 3 | 234 | 72 | ||
| NM_171477.7 | NP_741559.3 | 3 - 4 | 287 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001373047.2 transcripts.
Set Membership: |
|

mec-1
Genomic Map