Assay Details
Gene Symbol: smg-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171514.6 | NP_741600.1 | 1 - 2 | 149 | 127 | |
NM_171515.5 | NP_741601.1 | 1 - 2 | 147 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
