Assay Details
| Gene Symbol: pph-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269921.3 | NP_001256850.1 | 2 - 3 | 271 | 103 | |
| NM_001380870.1 | NP_001367002.1 | 7 - 8 | 1416 | 103 | ||
More Information
Set Membership: |
|

pph-5
Genomic Map