Assay Details
| Gene Symbol: unc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348588.4 | NP_001335561.1 | 27 - 28 | 5778 | 100 | |
| NM_001348589.4 | NP_001335563.1 | 27 - 28 | 5784 | 100 | ||
| NM_001348590.4 | NP_001335560.1 | 28 - 29 | 5832 | 100 | ||
| NM_001348591.4 | NP_001335562.1 | 28 - 29 | 5838 | 100 | ||
| NM_001348592.5 | NP_001335571.1 | 27 - 28 | 5973 | 100 | ||
| NM_001348593.4 | NP_001335573.1 | 27 - 28 | 5979 | 100 | ||
| NM_001348594.5 | NP_001335570.1 | 28 - 29 | 6027 | 100 | ||
| NM_001348595.4 | NP_001335572.1 | 28 - 29 | 6033 | 100 | ||
| NM_001348596.5 | NP_001335516.1 | 26 - 27 | 5565 | 100 | ||
| NM_001368494.4 | NP_001355464.1 | 33 - 34 | 6345 | 100 | ||
| NM_001368495.4 | NP_001355463.1 | 31 - 32 | 6189 | 100 | ||
| NM_001368496.4 | NP_001355462.1 | 31 - 32 | 6213 | 100 | ||
| NM_001368497.4 | NP_001355494.1 | 27 - 28 | 6112 | 100 | ||
| NM_171638.3 | NP_741732.2 | 27 - 28 | 5969 | 100 | ||
| GenBank mRNA | AY264781.1 | - | 27 - 28 | 5969 | 100 | |
More Information
Set Membership: |
|

unc-2
Genomic Map