Assay Details
| Gene Symbol: lgx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373298.3 | NP_001359928.1 | 11 - 12 | 1796 | 60 | |
| NM_001373299.2 | NP_001360723.1 | 11 | 1743 | 60 | ||
| NM_001373300.2 | NP_001360725.1 | 12 | 1887 | 60 | ||
| NM_001373301.2 | NP_001360721.1 | 12 | 1887 | 60 | ||
| NM_001373302.2 | NP_001360726.1 | 12 | 1896 | 60 | ||
| NM_001373303.2 | NP_001360722.1 | 12 | 1896 | 60 | ||
| NM_001373304.2 | NP_001360724.1 | 13 | 2040 | 60 | ||
| NM_001373305.2 | NP_001360720.1 | 13 | 2040 | 60 | ||
| NM_171690.6 | NP_741796.2 | 13 - 14 | 2095 | 60 | ||
More Information
Set Membership: |
|

lgx-1
Genomic Map