Assay Details
| Gene Symbol: lgx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373298.3 | NP_001359928.1 | 27 - 28 | 4302 | 80 | |
| NM_001373299.2 | NP_001360723.1 | 27 | 4552 | 80 | ||
| NM_001373300.2 | NP_001360725.1 | 28 | 4393 | 80 | ||
| NM_001373301.2 | NP_001360721.1 | 28 | 4696 | 80 | ||
| NM_001373302.2 | NP_001360726.1 | 28 | 4402 | 80 | ||
| NM_001373303.2 | NP_001360722.1 | 28 | 4705 | 80 | ||
| NM_001373304.2 | NP_001360724.1 | 29 | 4546 | 80 | ||
| NM_001373305.2 | NP_001360720.1 | 29 | 4849 | 80 | ||
More Information
Set Membership: |
|

lgx-1
Genomic Map