Assay Details
Gene Symbol: pak-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029206.3 | NP_001024377.1 | 2 - 3 | 170 | 76 | |
NM_001029207.4 | NP_001024378.1 | 2 - 3 | 168 | 76 | ||
GenBank mRNA | D83215.1 | - | 2 - 3 | 165 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
