Assay Details
| Gene Symbol: unc-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029730.6 | NP_001024901.1 | 20 - 21 | 3343 | 70 | |
| NM_001375064.2 | NP_001362128.1 | 21 | 3298 | 70 | ||
| NM_001375065.2 | NP_001362126.1 | 21 | 3298 | 70 | ||
| NM_001375066.2 | NP_001362127.1 | 21 | 3304 | 70 | ||
| NM_001375067.2 | NP_001362125.1 | 21 | 3304 | 70 | ||
| NM_171720.7 | NP_741831.1 | 19 - 20 | 3412 | 70 | ||
| GenBank mRNA | AF257062.1 | - | 19 - 20 | 3391 | 70 | |
More Information
Set Membership: |
|

unc-10
Genomic Map