Assay Details
| Gene Symbol: unc-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171720.7 | NP_741831.1 | 23 - 24 | 4178 | 74 | |
| GenBank mRNA | AF257062.1 | - | 23 - 24 | 4157 | 74 | |
More Information
Set Membership: |
|

unc-10
Genomic Map