Assay Details
| Gene Symbol: unc-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029730.6 | NP_001024901.1 | 25 - 26 | 4294 | 77 | |
| NM_001375064.2 | NP_001362128.1 | 25 | 3930 | 77 | ||
| NM_001375065.2 | NP_001362126.1 | 26 | 4245 | 77 | ||
| NM_001375066.2 | NP_001362127.1 | 25 | 3936 | 77 | ||
| NM_001375067.2 | NP_001362125.1 | 26 | 4251 | 77 | ||
| NM_171720.7 | NP_741831.1 | 24 - 25 | 4374 | 77 | ||
| GenBank mRNA | AF257062.1 | - | 24 - 25 | 4353 | 77 | |
More Information
Set Membership: |
|

unc-10
Genomic Map