Assay Details
| Gene Symbol: unc-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029730.6 | NP_001024901.1 | 8 - 9 | 1081 | 89 | |
| NM_001375064.2 | NP_001362128.1 | 8 | 1081 | 89 | ||
| NM_001375065.2 | NP_001362126.1 | 8 | 1081 | 89 | ||
| NM_001375066.2 | NP_001362127.1 | 8 | 1081 | 89 | ||
| NM_001375067.2 | NP_001362125.1 | 8 | 1081 | 89 | ||
| NM_171720.7 | NP_741831.1 | 8 - 9 | 1102 | 89 | ||
| GenBank mRNA | AF257062.1 | - | 8 - 9 | 1081 | 89 | |
More Information
Set Membership: |
|

unc-10
Genomic Map