Assay Details
| Gene Symbol: cca-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381028.1 | NP_001367692.1 | 21 - 22 | 3834 | 77 | |
| NM_001381029.1 | NP_001368577.1 | 21 - 22 | 3758 | 77 | ||
| NM_001381030.1 | NP_001367691.1 | 22 - 23 | 3957 | 77 | ||
| NM_001381031.2 | NP_001367690.1 | 21 - 22 | 3813 | 77 | ||
| NM_171733.5 | NP_741848.1 | 23 - 24 | 3899 | 77 | ||
| GenBank mRNA | AF368920.1 | - | 21 - 22 | 3762 | 77 | |
| AY313898.1 | - | 21 - 22 | 3813 | 77 | ||
| AY313899.1 | - | 22 - 23 | 3957 | 77 | ||
| AY313900.1 | - | 21 - 22 | 3813 | 77 | ||
| AY322480.1 | - | 21 - 22 | 3834 | 77 | ||
More Information
Set Membership: |
|

cca-1
Genomic Map